Product Details

SNP ID
rs201768155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43880880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTTTCCAGGACGCGAGTGGCC[A/G]GAGGGCTGTGGAGGGAGGAGAAGCC
Phenotype
MIM: 611589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPLA5 PubMed Links

Gene Details

Gene
PNPLA5
Gene Name
patatin like phospholipase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177675.1 996 Missense Mutation CCG,CTG P288L NP_001171146.1
NM_138814.3 996 Missense Mutation CCG,CTG P402L NP_620169.1
XM_011529959.2 996 Missense Mutation CCG,CTG P262L XP_011528261.1

View Full Product Details