Product Details

SNP ID
rs199507852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169735174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAATTTTCCATTGTCACCGAGCC[A/G]TTTTAGCTGCTTGCAGCAATTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL23 PubMed Links

Gene Details

Gene
KLHL23
Gene Name
kelch like family member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144711.5 404 Missense Mutation ATT,GTT I54V NP_653312.2
Gene
PHOSPHO2-KLHL23
Gene Name
PHOSPHO2-KLHL23 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199290.1 404 Missense Mutation ATT,GTT I54V NP_001186219.1

View Full Product Details