Product Details

SNP ID
rs199779856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27036553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCTCACCATCATGTGGGCCCTG[A/G]GTCAAGCAGGTTTTGCCAACCTCAC
Phenotype
MIM: 605788 MIM: 615301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPRE3 PubMed Links

Gene Details

Gene
MAPRE3
Gene Name
microtubule associated protein RP/EB family member 3
There are no transcripts associated with this gene.

Gene
TMEM214
Gene Name
transmembrane protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083590.1 870 Missense Mutation AGT,GGT S218G NP_001077059.1
NM_017727.4 870 Missense Mutation AGT,GGT S263G NP_060197.4
XM_005264381.3 870 Missense Mutation AGT,GGT S263G XP_005264438.1
XM_005264382.3 870 Missense Mutation AGT,GGT S263G XP_005264439.1
XM_005264383.3 870 Intron XP_005264440.1

View Full Product Details