Product Details

SNP ID
rs201559196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74505353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCTTACCTTGCCGAACCAGC[G/T]GGAGAGCCATATCTGCTTCATTGTC
Phenotype
MIM: 607164 MIM: 610231 MIM: 604240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LBX2 PubMed Links

Gene Details

Gene
LBX2
Gene Name
ladybird homeobox 2
There are no transcripts associated with this gene.

Gene
LBX2-AS1
Gene Name
LBX2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PCGF1
Gene Name
polycomb group ring finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032673.2 744 Missense Mutation NP_116062.2
Gene
TLX2
Gene Name
T-cell leukemia homeobox 2
There are no transcripts associated with this gene.

View Full Product Details