Product Details
- SNP ID
-
rs202002610
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:119146790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGATGGACCGCAGAATGTCTGGC[A/G]AGGCTGAGCGAAGAACTTCTGAGCA
- Phenotype
-
MIM: 608351
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf30
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10934483,rs9859242] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C3orf30
- Gene Name
- chromosome 3 open reading frame 30
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152539.2 |
641 |
Missense Mutation |
AAG,GAG |
K201E |
NP_689752.2 |
- Gene
- IGSF11
- Gene Name
- immunoglobulin superfamily member 11
View Full Product Details