Product Details

SNP ID
rs202088564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9839142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCACAGGGCAGAGCTGTAGTA[C/T]CATGTGCACCTGTAGTTGACTGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC4-TTLL3 PubMed Links

Gene Details

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
There are no transcripts associated with this gene.

Gene
LOC107986062
Gene Name
endogenous retrovirus group K member 9 Pol protein-like
There are no transcripts associated with this gene.

Gene
RPUSD3
Gene Name
RNA pseudouridylate synthase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142547.1 758 Missense Mutation NP_001136019.1
NM_173659.3 758 Missense Mutation NP_775930.2
XM_011533627.2 758 Intron XP_011531929.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
There are no transcripts associated with this gene.

View Full Product Details