Product Details

SNP ID
rs200752492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98799379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAGCAGCATCATCTTCAAAGG[A/T]TTTCTTTAAAAACATCACATTCCCC
Phenotype
MIM: 608698 MIM: 607156
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCBLD2 PubMed Links

Gene Details

Gene
DCBLD2
Gene Name
discoidin, CUB and LCCL domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080927.3 2400 Missense Mutation AAC,ATC N774I NP_563615.3
XM_011512419.2 2400 Missense Mutation AAC,ATC N698I XP_011510721.1
Gene
ST3GAL6
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details