Product Details

SNP ID
rs200111812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160677422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTGGGACTTGACTCAGCTGGG[A/T]AGTCTACTCTCCTTTATAAATTAAA
Phenotype
MIM: 614439
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL14 PubMed Links

Gene Details

Gene
ARL14
Gene Name
ADP ribosylation factor like GTPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025047.2 263 Nonsense Mutation AAG,TAG K26* NP_079323.1

View Full Product Details