Product Details
- SNP ID
-
rs200184954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138264131 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTAATTTTGATTATTCTTTGTAG[A/G]TGTTTGCACAGTTTATCCAGATCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC8
PubMed Links
Gene Details
- Gene
- ARMC8
- Gene Name
- armadillo repeat containing 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001267041.2 |
1520 |
Silent Mutation |
AGA,AGG |
R375R |
NP_001253970.1 |
NM_001267042.2 |
1520 |
Silent Mutation |
AGA,AGG |
R339R |
NP_001253971.1 |
NM_001282342.1 |
1520 |
Silent Mutation |
AGA,AGG |
R333R |
NP_001269271.1 |
NM_014154.4 |
1520 |
Intron |
|
|
NP_054873.2 |
NM_015396.5 |
1520 |
Silent Mutation |
AGA,AGG |
R392R |
NP_056211.2 |
NM_213654.3 |
1520 |
Intron |
|
|
NP_998819.1 |
XM_006713564.3 |
1520 |
Silent Mutation |
AGA,AGG |
R406R |
XP_006713627.1 |
XM_006713565.2 |
1520 |
Silent Mutation |
AGA,AGG |
R392R |
XP_006713628.1 |
XM_006713566.3 |
1520 |
Silent Mutation |
AGA,AGG |
R364R |
XP_006713629.1 |
XM_006713567.3 |
1520 |
Silent Mutation |
AGA,AGG |
R406R |
XP_006713630.1 |
XM_011512632.1 |
1520 |
Silent Mutation |
AGA,AGG |
R364R |
XP_011510934.1 |
XM_011512633.1 |
1520 |
Silent Mutation |
AGA,AGG |
R364R |
XP_011510935.1 |
XM_017006095.1 |
1520 |
Silent Mutation |
AGA,AGG |
R325R |
XP_016861584.1 |
XM_017006096.1 |
1520 |
Silent Mutation |
AGA,AGG |
R364R |
XP_016861585.1 |
XM_017006097.1 |
1520 |
Silent Mutation |
AGA,AGG |
R297R |
XP_016861586.1 |
XM_017006098.1 |
1520 |
Silent Mutation |
AGA,AGG |
R297R |
XP_016861587.1 |
XM_017006099.1 |
1520 |
Silent Mutation |
AGA,AGG |
R297R |
XP_016861588.1 |
XM_017006100.1 |
1520 |
Silent Mutation |
AGA,AGG |
R406R |
XP_016861589.1 |
XM_017006101.1 |
1520 |
Intron |
|
|
XP_016861590.1 |
- Gene
- NME9
- Gene Name
- NME/NM23 family member 9
View Full Product Details