Product Details

SNP ID
rs200376617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46021180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGTTTGGCCTCGCAGCTCCG[G/T]ATGATCTGGGTCCGAAACAGCGTCT
Phenotype
MIM: 600552
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
XCR1 PubMed Links

Gene Details

Gene
XCR1
Gene Name
X-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024644.1 1072 Silent Mutation NP_001019815.1
NM_005283.2 1072 Silent Mutation NP_005274.1
XM_017006196.1 1072 Silent Mutation XP_016861685.1
XM_017006197.1 1072 Silent Mutation XP_016861686.1
XM_017006198.1 1072 Silent Mutation XP_016861687.1
XM_017006199.1 1072 Silent Mutation XP_016861688.1
XM_017006200.1 1072 Silent Mutation XP_016861689.1
XM_017006201.1 1072 Silent Mutation XP_016861690.1

View Full Product Details