Product Details

SNP ID
rs200382005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48885532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTTCTCACTTAATCCATGCTT[G/T]ATTTTGCATTCCTCACAGGTAAGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OCIAD2 PubMed Links

Gene Details

Gene
OCIAD2
Gene Name
OCIA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014446.2 650 Silent Mutation ATA,ATC I139I NP_001014446.1
NM_001286774.1 650 Silent Mutation ATA,ATC I77I NP_001273703.1

View Full Product Details