Product Details

SNP ID
rs199812310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154235042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATCACCTGAAATAAAGACAG[A/G]CTCTTTTCTAACGTCAGCCAGGATG
Phenotype
MIM: 612486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCHS2 PubMed Links

Gene Details

Gene
DCHS2
Gene Name
dachsous cadherin-related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142552.1 5539 Intron NP_001136024.1
NM_017639.3 5539 Missense Mutation CCT,TCT P2749S NP_060109.2
XM_011532045.1 5539 Missense Mutation CCT,TCT P3204S XP_011530347.1
XM_011532046.2 5539 Missense Mutation CCT,TCT P2427S XP_011530348.1
XM_011532049.2 5539 Missense Mutation CCT,TCT P1539S XP_011530351.1
XM_017008320.1 5539 Intron XP_016863809.1

View Full Product Details