Product Details

SNP ID
rs202048868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:137521404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGCGGTAAGAGACGCTGGAA[C/T]ACACTGCTCATTTCCGAGAGCAGAG
Phenotype
MIM: 608287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH18 PubMed Links

Gene Details

Gene
PCDH18
Gene Name
protocadherin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300828.1 3443 Silent Mutation GTA,GTG V1010V NP_001287757.1
NM_019035.4 3443 Silent Mutation GTA,GTG V1011V NP_061908.1
XM_006714239.3 3443 Intron XP_006714302.1
XM_017008311.1 3443 UTR 3 XP_016863800.1

View Full Product Details