Product Details

SNP ID
rs201692437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56401337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGAAATCTACTTACCTTCGAA[C/T]ATACTGTCTGGTCTTGCAAAATAAA
Phenotype
MIM: 172450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506514 PubMed Links

Gene Details

Gene
LOC100506514
Gene Name
uncharacterized LOC100506514
There are no transcripts associated with this gene.

Gene
PPAT
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002703.4 Intron NP_002694.3
XM_011534387.1 Intron XP_011532689.1

View Full Product Details