Product Details

SNP ID
rs202245167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159916930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAATCCCGACCTGGACACCGGC[C/T]ACAACACATCAGCACCTGCCCACTG
Phenotype
MIM: 104220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA1B PubMed Links

Gene Details

Gene
ADRA1B
Gene Name
adrenoceptor alpha 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000679.3 198 Missense Mutation CAC,TAC H9Y NP_000670.1
XM_005265818.3 198 Missense Mutation CAC,TAC H9Y XP_005265875.1
XM_005265819.2 198 Missense Mutation CAC,TAC H9Y XP_005265876.1
XM_006714821.3 198 Missense Mutation CAC,TAC H9Y XP_006714884.1
XM_011534435.1 198 Missense Mutation CAC,TAC H9Y XP_011532737.1
XM_011534437.2 198 Missense Mutation CAC,TAC H9Y XP_011532739.1
XM_011534438.2 198 Missense Mutation CAC,TAC H9Y XP_011532740.1

View Full Product Details