Product Details

SNP ID
rs202030822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:7832016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGAAGACGTCAGCACTCCAACC[A/G]GCCTCTCCTGTTCCTGGTGAAAGTT
Phenotype
MIM: 103071
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY2 PubMed Links

Gene Details

Gene
ADCY2
Gene Name
adenylate cyclase 2
There are no transcripts associated with this gene.

Gene
C5orf49
Gene Name
chromosome 5 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001089584.2 747 Missense Mutation CCG,CTG P93L NP_001083053.1
XM_011513963.2 747 Missense Mutation CCG,CTG P61L XP_011512265.1

View Full Product Details