Product Details

SNP ID
rs201877962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149505580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCAATCAAATGTGTAGTCATA[C/T]TGATGGTTCAGGGTCCTGGAACACA
Phenotype
MIM: 600505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSNK1A1 PubMed Links

Gene Details

Gene
CSNK1A1
Gene Name
casein kinase 1 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025105.2 1461 Silent Mutation CAA,CAG Q319Q NP_001020276.1
NM_001271741.1 1461 Silent Mutation CAA,CAG Q291Q NP_001258670.1
NM_001271742.1 1461 Intron NP_001258671.1
NM_001892.5 1461 Silent Mutation CAA,CAG Q291Q NP_001883.4

View Full Product Details