Product Details
- SNP ID
-
rs200337257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:111092567 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTGGTGCGGTTCAGCGCGCTCAA[G/T]CGCCGGTTCTATGTAACAACCTGCG
- Phenotype
-
MIM: 609669
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR36
PubMed Links
Gene Details
- Gene
- WDR36
- Gene Name
- WD repeat domain 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139281.2 |
396 |
Missense Mutation |
AAG,AAT |
K93N |
NP_644810.1 |
View Full Product Details