Product Details

SNP ID
rs201603300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74769344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGTACTTTCCAATATGATTAAA[A/C]AGAAAAGAAAAGAGAAGGCGGTAAG
Phenotype
MIM: 615769 MIM: 606544 MIM: 612497
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM169A PubMed Links

Gene Details

Gene
FAM169A
Gene Name
family with sequence similarity 169 member A
There are no transcripts associated with this gene.

Gene
GFM2
Gene Name
G elongation factor mitochondrial 2
There are no transcripts associated with this gene.

Gene
NSA2
Gene Name
NSA2, ribosome biogenesis homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271665.1 692 Missense Mutation AAG,CAG K108Q NP_001258594.1
NM_014886.4 692 Missense Mutation AAG,CAG K108Q NP_055701.1
XM_011543098.1 692 Intron XP_011541400.1
XM_017008952.1 692 Intron XP_016864441.1
XM_017008953.1 692 Intron XP_016864442.1

View Full Product Details