Product Details

SNP ID
rs200893095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159917229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCAGCGGCCCTAGAGGTGCT[C/T]GGCTACTGGGTGCTGGGGCGGATCT
Phenotype
MIM: 104220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA1B PubMed Links

Gene Details

Gene
ADRA1B
Gene Name
adrenoceptor alpha 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000679.3 497 Silent Mutation CTC,CTT L108L NP_000670.1
XM_005265818.3 497 Silent Mutation CTC,CTT L108L XP_005265875.1
XM_005265819.2 497 Silent Mutation CTC,CTT L108L XP_005265876.1
XM_006714821.3 497 Silent Mutation CTC,CTT L108L XP_006714884.1
XM_011534435.1 497 Silent Mutation CTC,CTT L108L XP_011532737.1
XM_011534437.2 497 Silent Mutation CTC,CTT L108L XP_011532739.1
XM_011534438.2 497 Silent Mutation CTC,CTT L108L XP_011532740.1

View Full Product Details