Product Details

SNP ID
rs201232840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64774769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTAAAGCTTGCAGAAATTTT[A/G]TCCAACTTTGTTTGGAAGGTATGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CWC27 PubMed Links

Gene Details

Gene
CWC27
Gene Name
CWC27 spliceosome associated protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297644.1 350 Missense Mutation ATC,GTC I41V NP_001284573.1
NM_001297645.1 350 Missense Mutation ATC,GTC I41V NP_001284574.1
NM_001318000.1 350 Missense Mutation ATC,GTC I41V NP_001304929.1
NM_005869.3 350 Missense Mutation ATC,GTC I41V NP_005860.2
XM_011543095.2 350 Missense Mutation ATC,GTC I41V XP_011541397.2
XM_011543096.1 350 Missense Mutation ATC,GTC I41V XP_011541398.1
XM_017008943.1 350 Missense Mutation ATC,GTC I41V XP_016864432.1
Gene
SREK1IP1
Gene Name
SREK1 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details