Product Details

SNP ID
rs200828307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32857363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTGATGCCCAAGCCGTGGCC[A/G]ACATGGCCGCCTACCAGCTGGAGCT
Phenotype
MIM: 177045 MIM: 170260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMB9 PubMed Links

Gene Details

Gene
PSMB9
Gene Name
proteasome subunit beta 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002800.4 298 Missense Mutation AAC,GAC N77D NP_002791.1
Gene
TAP1
Gene Name
transporter 1, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

View Full Product Details