Product Details

SNP ID
rs201106154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43512518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGCCAGGGTGCCACAGAGGA[A/G]CAGCCGCTGTGTGGGGCCCACGGTC
Phenotype
MIM: 610060 MIM: 612658 MIM: 607845 MIM: 609775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
There are no transcripts associated with this gene.

Gene
POLR1C
Gene Name
RNA polymerase I subunit C
There are no transcripts associated with this gene.

Gene
TJAP1
Gene Name
tight junction associated protein 1
There are no transcripts associated with this gene.

Gene
XPO5
Gene Name
exportin 5
There are no transcripts associated with this gene.

Gene
YIPF3
Gene Name
Yip1 domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015388.3 1045 Missense Mutation CTC,TTC L276F NP_056203.2
XM_005248989.4 1045 Missense Mutation CTC,TTC L241F XP_005249046.1
XM_005248990.2 1045 Missense Mutation CTC,TTC L241F XP_005249047.1
XM_011514458.2 1045 Missense Mutation CTC,TTC L241F XP_011512760.1

View Full Product Details