Product Details

SNP ID
rs201427999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCCGGCGCCCAGTGGGCATCG[C/G]CGTCTATGGACACCCCTCCCACCTG
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 698 Missense Mutation GCC,GGC A117G NP_001025034.1
NM_001317342.1 698 Missense Mutation GCC,GGC A136G NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 698 Intron NP_001189479.1
NM_182552.4 698 Intron NP_872358.4
XM_011535682.2 698 Intron XP_011533984.1
XM_011535684.2 698 Intron XP_011533986.1
XM_011535685.2 698 Intron XP_011533987.1
XM_011535687.2 698 Intron XP_011533989.1
XM_011535688.2 698 Intron XP_011533990.1
XM_011535689.2 698 Intron XP_011533991.1
XM_011535690.2 698 Intron XP_011533992.1
XM_011535691.2 698 Intron XP_011533993.1
XM_011535692.2 698 Intron XP_011533994.1
XM_011535693.2 698 Intron XP_011533995.1
XM_011535694.2 698 Intron XP_011533996.1
XM_011535696.2 698 Intron XP_011533998.1
XM_011535697.2 698 Intron XP_011533999.1
XM_017010658.1 698 Intron XP_016866147.1
XM_017010659.1 698 Intron XP_016866148.1
XM_017010660.1 698 Intron XP_016866149.1
XM_017010661.1 698 Intron XP_016866150.1
XM_017010662.1 698 Intron XP_016866151.1
XM_017010663.1 698 Intron XP_016866152.1
XM_017010664.1 698 Intron XP_016866153.1
XM_017010665.1 698 Intron XP_016866154.1
XM_017010666.1 698 Intron XP_016866155.1
XM_017010667.1 698 Intron XP_016866156.1
XM_017010668.1 698 Intron XP_016866157.1
XM_017010669.1 698 Intron XP_016866158.1
XM_017010670.1 698 Intron XP_016866159.1
XM_017010671.1 698 Intron XP_016866160.1
XM_017010672.1 698 Intron XP_016866161.1
XM_017010673.1 698 Intron XP_016866162.1
XM_017010674.1 698 Intron XP_016866163.1
XM_017010675.1 698 Intron XP_016866164.1

View Full Product Details