Product Details

SNP ID
rs199632685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131950438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATGCTGCACAGGGGCATGCAG[C/T]CCACCGCCCCGTCCAGGCACGTGCA
Phenotype
MIM: 121009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTGF PubMed Links

Gene Details

Gene
CTGF
Gene Name
connective tissue growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001901.2 601 Missense Mutation GAC,GGC D132G NP_001892.1

View Full Product Details