Product Details

SNP ID
rs199824026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89083839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTATACACTTGTATGAGCAACC[A/G]AAGATAAACAGACAGAAAAGCAAAT
Phenotype
MIM: 606714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNRC1 PubMed Links

Gene Details

Gene
PNRC1
Gene Name
proline rich nuclear receptor coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006813.2 812 Silent Mutation CCA,CCG P209P NP_006804.1

View Full Product Details