Product Details

SNP ID
rs200180483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18236549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTCTTCATCTGTAGGGGGTTTC[C/T]TCAACTTTTTAATTAAAGGTTCATC
Phenotype
MIM: 125264
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEK PubMed Links

Gene Details

Gene
DEK
Gene Name
DEK proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134709.1 1041 Missense Mutation AAG,AGG K283R NP_001128181.1
NM_003472.3 1041 Missense Mutation AAG,AGG K317R NP_003463.1
XM_017011273.1 1041 Missense Mutation AAG,AGG K317R XP_016866762.1

View Full Product Details