Product Details

SNP ID
rs200584566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92450756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATCACTGCAGAATCAATCTTCT[A/G]CAAGGTAAGCTTTTGTAGAGTTACT
Phenotype
MIM: 614518
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATAD1 PubMed Links

Gene Details

Gene
GATAD1
Gene Name
GATA zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021167.4 713 Missense Mutation TAC,TGC Y144C NP_066990.3

View Full Product Details