Product Details
- SNP ID
-
rs200684386
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73769438 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGCCCAGGCTGGCTCCCGGGCC[G/T]GTGGAGCGCGGCGCGGAGTAGACGA
- Phenotype
-
MIM: 602910
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN3
PubMed Links
Gene Details
- Gene
- CLDN3
- Gene Name
- claudin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306.3 |
833 |
Silent Mutation |
ACA,ACC |
T204T |
NP_001297.1 |
View Full Product Details