Product Details

SNP ID
rs200608757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92448876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGACATATATTTAAATTGAAAA[A/T]TGTAAGATATTTGTGGACAGTGCCT
Phenotype
MIM: 614518
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GATAD1 PubMed Links

Gene Details

Gene
GATAD1
Gene Name
GATA zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021167.4 656 Missense Mutation AAT,ATT N125I NP_066990.3
Gene
LOC105375396
Gene Name
bax inhibitor 1-like
There are no transcripts associated with this gene.

View Full Product Details