Product Details

SNP ID
rs200575329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66633381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTGGCAGCCATGTTCAGTGGGC[A/G]GCACTACATCCCCACGGACTCCGAG
Phenotype
MIM: 611725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD7 PubMed Links

Gene Details

Gene
KCTD7
Gene Name
potassium channel tetramerization domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167961.2 435 Missense Mutation CAG,CGG Q84R NP_001161433.1
NM_153033.4 435 Missense Mutation CAG,CGG Q84R NP_694578.1

View Full Product Details