Product Details

SNP ID
rs201137330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107767796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATTAAACTTTGAAGTACTGTA[A/G]TCTTTCTTCATCAAAATATGCAAAA
Phenotype
MIM: 606872 MIM: 126650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLL1 PubMed Links

Gene Details

Gene
CBLL1
Gene Name
Cbl proto-oncogene like 1
There are no transcripts associated with this gene.

Gene
SLC26A3
Gene Name
solute carrier family 26 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000111.2 2386 Silent Mutation GAC,GAT D725D NP_000102.1
XM_011515867.2 2386 Silent Mutation GAC,GAT D725D XP_011514169.1

View Full Product Details