Product Details
- SNP ID
-
rs199684927
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44756468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGTCTGTGCCTTGGCAACAAAGC[A/G]CCACTCAGCCGGCTGGATCGCTGTC
- Phenotype
-
MIM: 611196
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZMIZ2
PubMed Links
Gene Details
- Gene
- ZMIZ2
- Gene Name
- zinc finger MIZ-type containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300959.1 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
NP_001287888.1 |
NM_031449.3 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
NP_113637.3 |
NM_174929.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
NP_777589.2 |
XM_005249866.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249923.1 |
XM_005249867.4 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249924.1 |
XM_005249868.4 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249925.1 |
XM_005249869.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249926.1 |
XM_005249870.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249927.1 |
XM_005249871.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249928.1 |
XM_005249872.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249929.1 |
XM_005249873.2 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_005249930.1 |
XM_011515566.1 |
1261 |
Intron |
|
|
XP_011513868.1 |
XM_017012674.1 |
1261 |
Missense Mutation |
ACC,GCC |
T32A |
XP_016868163.1 |
View Full Product Details