Product Details

SNP ID
rs3192029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94130638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACACTCACCAATCACCAGAA[T/G]GGTGGTCAGCAGTATACCAACTGCC
Phenotype
MIM: 603017
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH17 PubMed Links

Gene Details

Gene
CDH17
Gene Name
cadherin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144663.1 2512 Missense Mutation NP_001138135.1
NM_004063.3 2512 Missense Mutation NP_004054.3
XM_011516790.1 2512 Missense Mutation XP_011515092.1

View Full Product Details