Product Details
- SNP ID
-
rs200910539
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85245840 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCCGAGGGACCATGTCGAGGCT[C/T]AGCTGGGGATACCGCGAGCACAACG
- Phenotype
-
MIM: 611436
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CA13
PubMed Links
Gene Details
- Gene
- CA13
- Gene Name
- carbonic anhydrase 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198584.2 |
354 |
Silent Mutation |
CTC,CTT |
L4L |
NP_940986.1 |
View Full Product Details