Product Details

SNP ID
rs201026746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10607167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGTCATTTTCTGAGTCTTTCT[C/G]CCAGCAGTTGCCCCAAGAGGATGCT
Phenotype
MIM: 608581
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RP1L1 PubMed Links

Gene Details

Gene
RP1L1
Gene Name
retinitis pigmentosa 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178857.5 7160 Missense Mutation CAG,GAG Q2311E NP_849188.4

View Full Product Details