Product Details

SNP ID
rs202025875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6936068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGAAACTCACACCACCAATGAG[G/T]CAGTTCCCAACACGAAGTTCTGTTC
Phenotype
MIM: 601157 MIM: 600471
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFA4 PubMed Links

Gene Details

Gene
DEFA4
Gene Name
defensin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001925.2 371 Nonsense Mutation TGA,TGC *82C NP_001916.1
Gene
DEFA6
Gene Name
defensin alpha 6
There are no transcripts associated with this gene.

View Full Product Details