Product Details
- SNP ID
-
rs200377849
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:51820654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCTGCATCTCATCATTTATGAA[A/C]TTTCTAAGTGTGCGTCGAATGTAAA
- Phenotype
-
MIM: 615904
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PCMTD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35006940] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCMTD1
- Gene Name
- protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
- Gene
- PXDNL
- Gene Name
- peroxidasin like
There are no transcripts associated with this gene.
View Full Product Details