Product Details

SNP ID
rs200403049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10607182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTTTCTGCCAGCAGTTGCCCC[A/T]AGAGGATGCTCTGGAGGAGGAAGGG
Phenotype
MIM: 608581
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RP1L1 PubMed Links

Gene Details

Gene
RP1L1
Gene Name
retinitis pigmentosa 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178857.5 7145 Missense Mutation AGG,TGG R2306W NP_849188.4

View Full Product Details