Product Details

SNP ID
rs200393292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143309535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTCTCCACTGGCACCCTGAAC[C/T]GCTTGCACCTGCGGGAGGCAGCATC
Phenotype
MIM: 606387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOP1MT PubMed Links

Gene Details

Gene
TOP1MT
Gene Name
topoisomerase (DNA) I, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258446.1 1742 Missense Mutation NP_001245375.1
NM_001258447.1 1742 Missense Mutation NP_001245376.1
NM_052963.2 1742 Missense Mutation NP_443195.1
XM_005250781.3 1742 Intron XP_005250838.1
Gene
ZNF696
Gene Name
zinc finger protein 696
There are no transcripts associated with this gene.

View Full Product Details