Product Details

SNP ID
rs200181030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGCGCCACTCAGATGTACCTC[G/T]CCCTTGCCATGGGCTCCACTGAGTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 310 Missense Mutation GCC,TCC A104S NP_001004485.1

View Full Product Details