Product Details

SNP ID
rs201208279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87727660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGAATTGCCTCAGCTACTCT[A/G]ACATTTGATCACAGTTTAGAGGCAC
Phenotype
MIM: 116880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSL PubMed Links

Gene Details

Gene
CTSL
Gene Name
cathepsin L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257971.1 947 Silent Mutation CTA,CTG L19L NP_001244900.1
NM_001257972.1 947 Silent Mutation CTA,CTG L19L NP_001244901.1
NM_001257973.1 947 UTR 5 NP_001244902.1
NM_001912.4 947 Silent Mutation CTA,CTG L19L NP_001903.1
NM_145918.2 947 Silent Mutation CTA,CTG L19L NP_666023.1
XM_005251716.3 947 Silent Mutation CTA,CTG L19L XP_005251773.1
XM_011518263.1 947 Silent Mutation CTA,CTG L19L XP_011516565.1
XM_017014293.1 947 Silent Mutation CTA,CTG L19L XP_016869782.1

View Full Product Details