Product Details

SNP ID
rs200433931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133474027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCTGCGAGGTCCATGGTGAGG[C/G]CGGCGATCAGCACGGACAGGAGCCG
Phenotype
MIM: 613104 MIM: 606813
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CACFD1 PubMed Links

Gene Details

Gene
CACFD1
Gene Name
calcium channel flower domain containing 1
There are no transcripts associated with this gene.

Gene
SLC2A6
Gene Name
solute carrier family 2 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145099.1 1056 Missense Mutation GCC,GGC A330G NP_001138571.1
NM_017585.3 1056 Missense Mutation GCC,GGC A330G NP_060055.2
XM_011518189.2 1056 Missense Mutation GCC,GGC A124G XP_011516491.2
XM_017014236.1 1056 Intron XP_016869725.1
XM_017014237.1 1056 Missense Mutation GCC,GGC A86G XP_016869726.1
XM_017014238.1 1056 Missense Mutation GCC,GGC A86G XP_016869727.1

View Full Product Details