Product Details

SNP ID
rs200642380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87727722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGCGATGCACAACAGATTAT[A/G]CGGCATGGTTAGTGAAACTTCCCCA
Phenotype
MIM: 116880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSL PubMed Links

Gene Details

Gene
CTSL
Gene Name
cathepsin L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257971.1 1009 Missense Mutation TAC,TGC Y40C NP_001244900.1
NM_001257972.1 1009 Missense Mutation TAC,TGC Y40C NP_001244901.1
NM_001257973.1 1009 Intron NP_001244902.1
NM_001912.4 1009 Missense Mutation TAC,TGC Y40C NP_001903.1
NM_145918.2 1009 Missense Mutation TAC,TGC Y40C NP_666023.1
XM_005251716.3 1009 Missense Mutation TAC,TGC Y40C XP_005251773.1
XM_011518263.1 1009 Missense Mutation TAC,TGC Y40C XP_011516565.1
XM_017014293.1 1009 Missense Mutation TAC,TGC Y40C XP_016869782.1

View Full Product Details