Product Details

SNP ID
rs200931352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127925237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGGTATCCAGTTCCATCTGGC[A/G]GAGGAACCAGCTCCGCTGGGGCCCT
Phenotype
MIM: 603564 MIM: 612865 MIM: 606378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM2 PubMed Links

Gene Details

Gene
DPM2
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
There are no transcripts associated with this gene.

Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135219.1 1880 Missense Mutation CGC,TGC R263C NP_001128691.1
NM_173492.1 1880 Missense Mutation CGC,TGC R60C NP_775763.1
XM_017014279.1 1880 Missense Mutation CCG,CTG P308L XP_016869768.1
Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
There are no transcripts associated with this gene.

View Full Product Details