Product Details

SNP ID
rs199638778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137279855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGGCTTCTTTCCTGACCAGG[A/C]CCGCGCGGAGAACCTGGCCGCGCAG
Phenotype
MIM: 611180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NELFB PubMed Links

Gene Details

Gene
NELFB
Gene Name
negative elongation factor complex member B
There are no transcripts associated with this gene.

Gene
TOR4A
Gene Name
torsin family 4 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017723.2 1283 Missense Mutation GAC,GCC D389A NP_060193.2

View Full Product Details