Product Details

SNP ID
rs199971797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108862657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGACCGCCAAGACTGGGGATA[A/G]TCGCAAGGAGACATTCGTGGGGCAG
Phenotype
MIM: 604303 MIM: 604304 MIM: 603722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL7A PubMed Links

Gene Details

Gene
ACTL7A
Gene Name
actin like 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006687.3 430 Missense Mutation AAT,AGT N112S NP_006678.1
Gene
ACTL7B
Gene Name
actin like 7B
There are no transcripts associated with this gene.

Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
There are no transcripts associated with this gene.

View Full Product Details