Product Details

SNP ID
rs202068838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91220880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCCTGGTTCTGTTCCAGAAC[A/G]TGGCTGATTAAGCCCACTGCTTTGG
Phenotype
MIM: 600529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUH PubMed Links

Gene Details

Gene
AUH
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306190.1 872 Silent Mutation CAC,CAT H227H NP_001293119.1
NM_001698.2 872 Silent Mutation CAC,CAT H256H NP_001689.1
XM_005252066.3 872 Silent Mutation CAC,CAT H266H XP_005252123.1
XM_005252067.4 872 Silent Mutation CAC,CAT H266H XP_005252124.1
XM_005252069.4 872 Silent Mutation CAC,CAT H266H XP_005252126.1
XM_005252072.2 872 Intron XP_005252129.1
XM_006717150.3 872 Silent Mutation CAC,CAT H237H XP_006717213.1
XM_011518800.2 872 Intron XP_011517102.1
XM_011518802.2 872 Silent Mutation CAC,CAT H147H XP_011517104.1
XM_011518803.2 872 Intron XP_011517105.1
XM_011518804.2 872 Intron XP_011517106.1
XM_017014849.1 872 Silent Mutation CAC,CAT H256H XP_016870338.1
XM_017014850.1 872 Silent Mutation CAC,CAT H147H XP_016870339.1
XM_017014851.1 872 Silent Mutation CAC,CAT H147H XP_016870340.1

View Full Product Details