Product Details

SNP ID
rs202128661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122918795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTCATTGGCATTATCAAAGCT[A/G]TTTTTGCCATGCAGCTGTTTAAGGT
Phenotype
MIM: 605976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB26 PubMed Links

Gene Details

Gene
ZBTB26
Gene Name
zinc finger and BTB domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304363.1 1223 Silent Mutation AAC,AAT N380N NP_001291292.1
NM_001304364.1 1223 Silent Mutation AAC,AAT N380N NP_001291293.1
NM_020924.3 1223 Silent Mutation AAC,AAT N380N NP_065975.1
Gene
ZBTB6
Gene Name
zinc finger and BTB domain containing 6
There are no transcripts associated with this gene.

View Full Product Details