Product Details

SNP ID
rs199861269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:116461911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCCCGAGAGAGGTCGTAGACTG[A/C]AAGATTGTTGTCTGTATTGCTGTTA
Phenotype
MIM: 300625 MIM: 300444
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CT83 PubMed Links

Gene Details

Gene
CT83
Gene Name
cancer/testis antigen 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017978.3 Intron NP_001017978.1
Gene
SLC6A14
Gene Name
solute carrier family 6 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007231.4 Intron NP_009162.1

View Full Product Details